siRNA Details
Gene Symbol: Gm7715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131765.1 | - | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707175.1 | - | 1 | 330 | Chr. 14 - 32513286 - 32513752 [-] on Build GRCm38 |
