siRNA Details
Gene Symbol: 4631402F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014506.1 | - | 1 | 369 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713919.1 | - | 1 | 369 | Chr. 17 - 65871467 - 65872418 [-] on Build GRCm38 |
