siRNA Details
Gene Symbol: Pdzd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088487.1 | - | 3370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY762603.1 | - | 1 | 355 | Chr. 19 - 59293231 - 59293890 [-] on Build GRCm38 |
