siRNA Details
Gene Symbol: Gm18409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038018.1 | 1 | 123 | |||
GenBank mRNA | AK039365.1 | - | 1 | 124 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633284.1 | - | 1 | 121 | Chr. 10 - 99640603 - 99641918 [-] on Build GRCm38 |
