siRNA Details
Gene Symbol: Tubb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013439.1 | - | 438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ230453.1 | - | 1 | 278 | Chr. 2 - 174458652 - 174459379 [+] on Build GRCm38 |
