siRNA Details
Gene Symbol: Gm31314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC050984.1 | - | 2 | 224 | ||
BC100494.1 | - | 2 | 245 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm31314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC050984.1 | - | 2 | 224 | ||
BC100494.1 | - | 2 | 245 |