siRNA Details
Gene Symbol: A930026C06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020892.1 | - | 1 | 574 | ||
AK048940.1 | - | 1 | 576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650259.1 | - | 1 | 571 | Chr. 7 - 107517543 - 107518158 [+] on Build GRCm38 |
