siRNA Details
Gene Symbol: Ube4bos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020898.1 | - | 539 | |||
BC024827.1 | - | 3 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742265.1 | - | 3 | 539 | Chr. 4 - 149347885 - 149364656 [+] on Build GRCm38 |
