siRNA Details
Gene Symbol: AW822252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110489.1 | 5, 6 | 772 | |||
GenBank mRNA | AK082553.1 | - | 5, 6 | 773 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB389450.2 | - | 3, 4 | 308 | Chr. X - 53696922 - 53715404 [-] on Build GRCm38 |
