siRNA Details
Gene Symbol: AW822252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110489.1 | 3 | 554 | |||
GenBank mRNA | AK082553.1 | - | 3 | 555 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE943729.1 | - | 2 | 174 | Chr. X - 53713358 - 53716480 [-] on Build GRCm38 |
