siRNA Details
Gene Symbol: 2310058D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009977.1 | - | 2 | 443 | ||
AK080053.1 | - | 2 | 462 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310058D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009977.1 | - | 2 | 443 | ||
AK080053.1 | - | 2 | 462 |