siRNA Details
Gene Symbol: 2310058D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA864041.1 | - | 1 | 94 | ||
AK009977.1 | - | 2 | 584 | |||
AK080053.1 | - | 2 | 603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709547.1 | - | 2 | 579 | Chr. 11 - 58968268 - 58978026 [-] on Build GRCm38 |
