siRNA Details
Gene Symbol: 2610034E01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011660.1 | - | 1 | 847 | ||
AK142273.1 | - | 1 | 645 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV075293.1 | - | 482 | Chr. - - [-] on Build |
