siRNA Details
Gene Symbol: 4930435M08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015326.1 | - | 2 | 1321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB384048.2 | - | 1 | 278 | Chr. 14 - 118046345 - 118046863 [+] on Build GRCm38 |
