siRNA Details
Gene Symbol: 4930588G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016366.1 | - | 3 | 685 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716013.1 | - | 3 | 685 | Chr. 7 - 120670303 - 120675017 [+] on Build GRCm38 |
