siRNA Details
Gene Symbol: Gm32442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135813.1 | - | 2 | 203 | ||
AK135963.1 | - | 2 | 220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719762.1 | - | 2 | 203 | Chr. 12 - 5808234 - 5856061 [+] on Build GRCm38 |
