siRNA Details
Gene Symbol: Gm11267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049966.1 | - | 2 | 111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB652897.1 | - | 2 | 111 | Chr. 4 - 82585494 - 82587319 [-] on Build GRCm38 |
