siRNA Details
Gene Symbol: Teddm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017892.1 | - | 858 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV312559.1 | - | 1 | 141 | Chr. 1 - 153899313 - 153899512 [-] on Build GRCm38 |
