siRNA Details
Gene Symbol: Gm20751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046022.1 | 3 | 820 | |||
GenBank mRNA | AK038993.1 | - | 3 | 821 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB180107.2 | - | 2 | 181 | Chr. 13 - 43718027 - 43719033 [+] on Build GRCm38 |
