siRNA Details
Gene Symbol: Gm20751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046022.1 | 1 | 597 | |||
GenBank mRNA | AK038993.1 | - | 1 | 598 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX207898.1 | - | 1 | 487 | Chr. 13 - 43716901 - 43718271 [+] on Build GRCm38 |
