siRNA Details
Gene Symbol: Gm16223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035725.1 | - | 391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB628355.1 | - | 3 | 391 | Chr. 5 - 42067960 - 42214930 [+] on Build GRCm38 |
