siRNA Details
Gene Symbol: Gm35147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035726.1 | - | 3, 4 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB628358.1 | - | 3, 4 | 486 | Chr. 7 - 132463523 - 132466661 [-] on Build GRCm38 |
