siRNA Details
Gene Symbol: Gm26515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131823.1 | - | 1 | 147 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707603.1 | - | 1 | 142 | Chr. 5 - 110779886 - 110780861 [+] on Build GRCm38 |
