siRNA Details
Gene Symbol: 5830454J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018012.1 | - | 1 | 95 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717943.1 | - | 1 | 93 | Chr. 17 - 28695103 - 28695753 [+] on Build GRCm38 |
