siRNA Details
Gene Symbol: 4930519P11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015848.1 | - | 1 | 1406 | ||
AK053350.1 | - | 1 | 1361 | |||
AV259580.2 | - | 1 | 402 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB538328.2 | - | 1 | 473 | Chr. 2 - 154611836 - 154612505 [-] on Build GRCm38 |
