siRNA Details
Gene Symbol: Gm20757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046027.1 | 9 | 1285 | |||
GenBank mRNA | AK053604.1 | - | 9 | 1286 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB541330.2 | - | 3 | 371 | Chr. 10 - 92171301 - 92257192 [-] on Build GRCm38 |
