siRNA Details
Gene Symbol: Gm20757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046027.1 | 2, 3 | 293 | |||
GenBank mRNA | AK053604.1 | - | 2, 3 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB665235.1 | - | 2, 3 | 294 | Chr. 10 - 92269329 - 92375373 [-] on Build GRCm38 |
