siRNA Details
Gene Symbol: 4930556C24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040506.1 | 4 | 462 | |||
GenBank mRNA | AK016142.1 | - | 4 | 463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715748.1 | - | 4 | 463 | Chr. 16 - 85819211 - 85826784 [+] on Build GRCm38 |
