siRNA Details
Gene Symbol: 4930556G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040655.1 | 5 | 469 | |||
GenBank mRNA | AK016143.1 | - | 5 | 470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715749.1 | - | 470 | Chr. - - [-] on Build |
