siRNA Details
Gene Symbol: 4930556J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045714.1 | 3 | 631 | |||
GenBank mRNA | AK016147.1 | - | 3 | 632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU945712.1 | - | 2 | 302 | Chr. 14 - 61493334 - 61500377 [+] on Build GRCm38 |
