siRNA Details
Gene Symbol: 5730411F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017546.1 | - | 1 | 757 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX517665.1 | - | 1 | 78 | Chr. 18 - 61535296 - 61535783 [-] on Build GRCm38 |
