siRNA Details
Gene Symbol: 1700022C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006235.1 | - | 1 | 748 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA451819.1 | - | 1 | 497 | Chr. 19 - 59346158 - 59346874 [+] on Build GRCm38 |
