siRNA Details
Gene Symbol: 9130223C08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020287.1 | - | 1 | 142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720865.1 | - | 1 | 142 | Chr. 4 - 53572440 - 53573037 [+] on Build GRCm38 |
