siRNA Details
Gene Symbol: 2310003N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009126.1 | - | 2 | 309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708964.1 | - | 2 | 313 | Chr. 9 - 50232523 - 50233182 [-] on Build GRCm38 |
