siRNA Details
Gene Symbol: Gm16226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160425.1 | - | 336 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707211.1 | - | 3 | 334 | Chr. 5 - 99682441 - 99715770 [+] on Build GRCm38 |
