siRNA Details
Gene Symbol: 6530403M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018320.1 | - | 1 | 612 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719216.1 | - | 1 | 612 | Chr. X - 56381199 - 56382113 [+] on Build GRCm38 |
