siRNA Details
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031725.1 | - | 1 | 1157 | ||
AK047638.1 | - | 1 | 4328 | |||
AK081408.1 | - | 1 | 1119 | |||
BC060127.1 | - | 1 | 3865 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE957170.1 | - | 1 | 508 | Chr. 4 - 126334291 - 126334808 [-] on Build GRCm38 |
