siRNA Details
Gene Symbol: 4930458D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015484.1 | - | 1 | 651 | ||
AK036594.1 | - | 1 | 1707 | |||
AK133807.1 | - | 1 | 643 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB028846.2 | - | 1 | 466 | Chr. 5 - 100841632 - 100845859 [-] on Build GRCm38 |
