siRNA Details
Gene Symbol: Gm36642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086666.1 | - | 6 | 1664 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB528091.2 | - | 3 | 642 | Chr. 15 - 25528879 - 25537005 [-] on Build GRCm38 |
