siRNA Details
Gene Symbol: Gm31063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081945.1 | - | 29 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649741.1 | - | 1 | 29 | Chr. 12 - 65187867 - 65213501 [-] on Build GRCm38 |
