siRNA Details
Gene Symbol: Gm31063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081945.1 | - | 534 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB377327.2 | - | 4 | 572 | Chr. 12 - 65187872 - 65213501 [-] on Build GRCm38 |
