siRNA Details
Gene Symbol: Gm32061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142799.1 | - | 3 | 606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB662750.1 | - | 3 | 603 | Chr. 7 - 62340638 - 62423244 [-] on Build GRCm38 |
