siRNA Details
Gene Symbol: Gm6846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049156.1 | - | 270 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB651697.1 | - | 1, 2 | 270 | Chr. 10 - 21834818 - 21844851 [-] on Build GRCm38 |
