siRNA Details
Gene Symbol: 4921508D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045802.1 | 3 | 363 | |||
GenBank mRNA | AK014842.1 | - | 3 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714363.1 | - | 3 | 365 | Chr. 2 - 132424752 - 132452357 [+] on Build GRCm38 |
