siRNA Details
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080872.1 | - | 1 | 931 | ||
AK154419.1 | - | 1 | 928 | |||
AK170106.1 | - | 1 | 759 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080872.1 | - | 1 | 931 | ||
AK154419.1 | - | 1 | 928 | |||
AK170106.1 | - | 1 | 759 |