siRNA Details
Gene Symbol: Gm33533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158561.1 | - | 2 | 377 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY239630.1 | - | 2 | 377 | Chr. 1 - 36740241 - 36744792 [-] on Build GRCm38 |
