siRNA Details
Gene Symbol: Gm38407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110487.1 | 2, 3 | 386 | |||
GenBank mRNA | AK029853.1 | - | 2, 3 | 388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616152.1 | - | 2, 3 | 388 | Chr. 12 - 10446388 - 10467795 [+] on Build GRCm38 |
