siRNA Details
Gene Symbol: 3110040K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014159.1 | - | 1 | 247 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713134.1 | - | 1 | 247 | Chr. 2 - 158411515 - 158412002 [+] on Build GRCm38 |
