siRNA Details
Gene Symbol: 1700054K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018860.1 | - | 1 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706768.1 | - | 1 | 341 | Chr. 1 - 92572369 - 92572749 [-] on Build GRCm38 |
