siRNA Details
Gene Symbol: 2310057M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014156.1 | - | 1 | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713132.1 | - | 1 | 364 | Chr. 7 - 131355166 - 131355742 [-] on Build GRCm38 |
